Learn More
Yttrium Aluminum lump, 99.9% (metals basis excluding Ca), Ca 1% max, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 012949.14
62.26 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.02500kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Yttrium Aluminum lump | |
(metals basis excluding Ca), Ca 1% max. | |
25 g | |
Lump |
Y:Al; 87:12 wt% | |
MFCD01074900 | |
99.9% |
RUO – Research Use Only