missing translation for 'onlineSavingsMsg'
Learn More

Yttrium Aluminum lump, 99.9% (metals basis excluding Ca), Ca 1% max, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 012949.22

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11312038

  • £303.00 / 100g
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Yttrium Aluminum lump
(metals basis excluding Ca), Ca 1% max.
100 g
Lump
Y:Al; 87:12 wt%
MFCD01074900
99.9%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only