missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ytterbium(III) oxalate hydrate, REacton™, 99.9% (REO)

Brand:  Alfa Aesar™ 014573.18

Code : NEW

Additional Details : CAS Number : 58176-74-2 Weight : 0.05000kg Transport : UN number : 3288 Chem class : 1 Pack group : III

Quantity 50g

 View more versions of this product

Product Code. 11392518

  • £122.32 / 50g


Estimated Shipment date
from Supplier 19-02-2018
Add to basket

Specifications

Chemical Identifiers

Ytterbium(III) oxalate hydrate
Reacton™
610.14 (Anhydrous)
UN3288
58176-74-2
Yb2(C2O4)3·xH2O
MFCD00050806

Specifications

Ytterbium(III) oxalate hydrate
Reacton™
99.9%
Yb2(C2O4)3·xH2O
MFCD00050806
50g
58176-74-2
Crystalline
(REO)
610.14 (Anhydrous)
UN3288
Videos
SDS