missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ytterbium(III) nitrate hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
53.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
Ytterbium(III) nitrate hydrate, is used as a precursor of nanoscale coatings of carbon composites. Source of yttrium used to make yttrium-based surfactant mesophases which are promising as adsorbing agents as well as for optically functional species.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Physical Form | Crystalline Aggregates |
Quantity | 10 g |
UN Number | UN1477 |
Merck Index | 14,10105 |
Solubility Information | Soluble in water. |
Formula Weight | 359.05 (Anhydrous) |
Percent Purity | 99.99% |
Grade | Reacton™ |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'