missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) isopropoxide, Thermo Scientific Chemicals

Catalog Number p-7050846
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
6742-69-4
C9H24O3Yb
353.342
MFCD00145532
IVGMSSQGEVYWID-UHFFFAOYSA-N
propan-2-ol; ytterbium, tris isopropanolato ytterbium
50912003
propan-2-ol;ytterbium
CC(C)O.CC(C)O.CC(C)O.[Yb]
This item is not returnable. View return policy
6742-69-4
C9H24O3Yb
353.342
MFCD00145532
IVGMSSQGEVYWID-UHFFFAOYSA-N
propan-2-ol; ytterbium, tris isopropanolato ytterbium
50912003
propan-2-ol;ytterbium
CC(C)O.CC(C)O.CC(C)O.[Yb]

Yttrium(III) tris(isopropoxide) is a catalyst for stereoselective conjugate addition reactions, ring-opening polymerization, generation of reactive enolates, allylic substitution and cyano-phosphorylation of enones and synthesis of nanocomposites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Melting Point 200°C (decomposition)
Boiling Point 190°C to 200°C (0.2 mmHg)(sublimation)
Odor Odorless
Packaging Ampule
Linear Formula Yb[OCH(CH3)2]3
Quantity 1 g
UN Number UN2925
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water. Soluble in hexane,toluene,and THF.
Formula Weight 350.31
Physical Form Powder
Chemical Name or Material Ytterbium(III) isopropoxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'