missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) bromide hydrate, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7049514
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
15163-03-8
MFCD00049613
ytterbium bromide hydrate, acmc-20aj9h, ytterbium 3+ bromide-water 1/3/1
This item is not returnable. View return policy
15163-03-8
MFCD00049613
ytterbium bromide hydrate, acmc-20aj9h, ytterbium 3+ bromide-water 1/3/1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 300°C
Color White
Physical Form Crystalline
Quantity 10 g
Solubility Information Soluble in water.
Formula Weight 412.77 (Anhydrous)
Percent Purity 99.9%
Sensitivity Very Hygroscopic
Assay (REO)
Chemical Name or Material Ytterbium(III) bromide hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'