missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium foil, 0.3mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7048402
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
7440-64-4
Yb
173.05
MFCD00011286
NAWDYIZEMPQZHO-UHFFFAOYSA-N
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
23992
CHEBI:33381
ytterbium
[Yb]
This item is not returnable. View return policy
7440-64-4
Yb
173.05
MFCD00011286
NAWDYIZEMPQZHO-UHFFFAOYSA-N
unii-mnq4o4wsi1, hydride, mnq4o4wsi1, foil, 3n, yterbio, ingot, powder, ii ion, chips, 3n, ingot, 3n
23992
CHEBI:33381
ytterbium
[Yb]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Quantity 25 x 25 mm
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈1.3g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Ytterbium foil, 0.3mm (0.01 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'