missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ytterbium fluoride oxide, 99.99% (REO)

Brand:  Alfa Aesar™ 041999.04

Code : NEW

Additional Details : CAS Number : 15587-02-7 Weight : 0.00200kg Transport : UN number : 3288 Chem class : 1 Pack group : III

Quantity 2g

 View more versions of this product

Product Code. 11335007

  • £24.10 / 2g


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Specifications

Chemical Identifiers

Ytterbium fluoride oxide
208.04
UN3288
15587-02-7
MFCD00467991
Ytterbium oxyfluoride

Specifications

Ytterbium fluoride oxide
Crystalline
(REO)
MFCD00467991
Ytterbium oxyfluoride
2g
15587-02-7
99.99%
208.04
UN3288
Odorless
Videos
SDS