Learn More
YSI™ pH Probes for Pro Series Portable Meters
Choose from lab-grade and field models.
Brand: YSI™ 605101
Code : 04
Additional Details : Weight : 0.02700kg
Description
Easy user-replaceable sensor allows for quick sensor replacements, if needed. No tools necessary. One-year warranty. Measure pH quickly and easily with the 1001 sensor. The 605101 pH Sensor can be used with the YSI Professional Plus Multiparameter Handheld Instrument. Requires Cable/Probe Assembly.
- For use with Pro Series Portable Meters
- Without cable
- Requires cable/probe assembly
Specifications
pH Probe | |
pH | |
Pro series meter | |
Without cable |