missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YSI™ Dip-Type Glass Micro Conductivity Cells
Brand: YSI™ 603253
Code : 04
Additional Details : Weight : 0.01000kg
Specifications
0.1 cm-1 | |
7-pin Mini-DIN | |
13 mm | |
48 in. | |
1.2 m | |
Micro Conductivity |
3200 Meter | |
0.5 in. | |
7 in. | |
431.8 mm | |
Pyrex 7740 Glass | |
Yes |