Learn More
YSI™ Carrying Cases and Harness for Model 556 Multi-Probe System
Small case accommodates instrument and 4m cable
£34.40
Product Code | Brand | Type | For Use With (Application) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | For Use With (Application) | Price | Quantity & Availability | ||||
12734556
|
YSI™
655227 |
Hands-free Harness | Hands free field measurements |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Carrying Cases and Harness. Hard-sided case is unbreakable, water- and air-tight and corrosion-proof. Small case accommodates instrument and 4m cable. Form-fitted case with clear vinyl window fits snugly over the instrument—allows operation and viewing while protecting instrument in the field. Comes with shoulder strap, belt-loop strap and hand strap. Harness permits hands-free maneuvering and one-handed operation of the instrument while in the field.