missing translation for 'onlineSavingsMsg'
Learn More

YSI™ Carrying Case for Pro Series Instruments

Brand:  YSI™ 603074

Code : 03

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11957127

  • £285.00 / Each
Estimated Shipment: 17-06-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Water quality testing
Pro series water quality testing instruments
520.7 mm
Carrying Case for Pro Series Instruments
20.5 in.
Hard sided carrying case
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers