missing translation for 'onlineSavingsMsg'
Learn More

YM 230888, Tocris Bioscience™

Selective mGlu1 antagonist

Brand:  Tocris (Bio-Techne) 298610

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13462194

  • £183.48 / 10mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Selective mGlu1 antagonist (Ki = 13 nM). Inhibits mGlu1-mediated inositol phosphate production in rat cerebellar granule cells (IC50 = 13 nM). Displays antinociceptive and analgesic effects in vitro; exhibits no significant sedative effect on locomotor activity.
Specifications

Chemical Identifiers

446257-23-4
360.52
4-cycloheptylamino-n-2r-tetrahydro-2-furanyl methyl-thieno 2,3-d pyrimidine-6-methanamine, n-cycloheptyl-6-2r-tetrahydrofuran-2-ylmethyl amino methyl thieno 2,3-d pyrimidin-4-amine
N-cycloheptyl-6-[[[(2R)-oxolan-2-yl]methylamino]methyl]thieno[2,3-d]pyrimidin-4-amine
C19H28N4OS
SASNRPJKZVJWQW-OAHLLOKOSA-N
11682007
C1CCCC(CC1)NC2=C3C=C(SC3=NC=N2)CNCC4CCCO4

Specifications

YM 230888
446257-23-4
4-cycloheptylamino-n-2r-tetrahydro-2-furanyl methyl-thieno 2,3-d pyrimidine-6-methanamine, n-cycloheptyl-6-2r-tetrahydrofuran-2-ylmethyl amino methyl thieno 2,3-d pyrimidin-4-amine
C1CCCC(CC1)NC2=C3C=C(SC3=NC=N2)CNCC4CCCO4
360.52
360.52
10mg
C19H28N4OS
SASNRPJKZVJWQW-OAHLLOKOSA-N
N-cycloheptyl-6-[[[(2R)-oxolan-2-yl]methylamino]methyl]thieno[2,3-d]pyrimidin-4-amine
11682007
>99%
Videos