Learn More
YK 4-279, Tocris Bioscience™
Inhibitor of RNA helicase A (RHA)
POA
Description
Description
Inhibitor of RNA Helicase A (RHA) binding to the oncogenic transciption factor EWS-FLI1. Inhibits Ewing's sarcoma family tumor (ESFT) cell growth; induces apoptosis.