missing translation for 'onlineSavingsMsg'
Learn More

YK 4-279, Tocris Bioscience™

Inhibitor of RNA helicase A (RHA)

Brand:  Tocris (Bio-Techne) 406750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13444074

  • £949.62 / 50mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of RNA Helicase A (RHA) binding to the oncogenic transciption factor EWS-FLI1. Inhibits Ewing's sarcoma family tumor (ESFT) cell growth; induces apoptosis.
Specifications

Chemical Identifiers

1037184-44-3
366.194
4,7-dichloro-3-hydroxy-3-2-4-methoxyphenyl-2-oxoethyl indolin-2-one, 4,7-dichloro-1,3-dihydro-3-hydroxy-3-2-4-methoxyphenyl-2-oxoethyl-2h-indol-2-one, 4,7-dichloro-3-hydroxy-3-2-4-methoxyphenyl-2-oxoethyl-1h-indol-2-one, 4,7-dichloro-3-hydroxy-3-2-4-methoxyphenyl-2-oxoethyl indolin-2-one yk-4-279
4,7-dichloro-3-hydroxy-3-[2-(4-methoxyphenyl)-2-oxoethyl]-1H-indol-2-one
C17H13Cl2NO4
HLXSCTYHLQHQDJ-UHFFFAOYSA-N
44632017
COC1=CC=C(C=C1)C(=O)CC2(C3=C(C=CC(=C3NC2=O)Cl)Cl)O

Specifications

50mg
1037184-44-3
4,7-dichloro-3-hydroxy-3-2-4-methoxyphenyl-2-oxoethyl indolin-2-one, 4,7-dichloro-1,3-dihydro-3-hydroxy-3-2-4-methoxyphenyl-2-oxoethyl-2h-indol-2-one, 4,7-dichloro-3-hydroxy-3-2-4-methoxyphenyl-2-oxoethyl-1h-indol-2-one, 4,7-dichloro-3-hydroxy-3-2-4-methoxyphenyl-2-oxoethyl indolin-2-one yk-4-279
COC1=CC=C(C=C1)C(=O)CC2(C3=C(C=CC(=C3NC2=O)Cl)Cl)O
366.194
366.2
YK 4-279
C17H13Cl2NO4
HLXSCTYHLQHQDJ-UHFFFAOYSA-N
4,7-dichloro-3-hydroxy-3-[2-(4-methoxyphenyl)-2-oxoethyl]-1H-indol-2-one
44632017
>99%
Videos
Documents