missing translation for 'onlineSavingsMsg'
Learn More

YIL 781, Tocris Bioscience™

Ghrelin receptor (GHS-R1a) antagonist

Brand:  Tocris (Bio-Techne) 395950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13463884

  • £702.68 / 50mg


Estimated Shipment
04-04-2019
Add to basket

Description

Description

Ghrelin receptor antagonist (GHS-R1a) (Ki = 17 nM). Displays no significant affinity for the motilin receptor (Ki = 6μM). Blocks the effects of ghrelin on insulin secretion both in vivo and in vitro. Improves glucose homeostasis in vivo.
Specifications

Chemical Identifiers

875258-85-8
445.963
6-4-fluorophenoxy-3-3s-1-isopropylpiperidin-3-yl methyl-2-methylquinazolin-4-one hydrochloride, yil-781 hydrochloride hplc, 6-4-fluorophenoxy-2-methyl-3-3s-1-1-methylethyl-3-piperidinyl methyl-4 3h-quinazolinone
6-(4-fluorophenoxy)-2-methyl-3-[[(3S)-1-propan-2-ylpiperidin-3-yl]methyl]quinazolin-4-one;hydrochloride
C24H29ClFN3O2
QQGRMONBDBMLSP-FERBBOLQSA-N
74889155
CC1=NC2=C(C=C(C=C2)OC3=CC=C(C=C3)F)C(=O)N1CC4CCCN(C4)C(C)C.Cl

Specifications

YIL 781
875258-85-8
6-4-fluorophenoxy-3-3s-1-isopropylpiperidin-3-yl methyl-2-methylquinazolin-4-one hydrochloride, yil-781 hydrochloride hplc, 6-4-fluorophenoxy-2-methyl-3-3s-1-1-methylethyl-3-piperidinyl methyl-4 3h-quinazolinone
CC1=NC2=C(C=C(C=C2)OC3=CC=C(C=C3)F)C(=O)N1CC4CCCN(C4)C(C)C.Cl
445.963
445.96
50mg
C24H29ClFN3O2
QQGRMONBDBMLSP-FERBBOLQSA-N
6-(4-fluorophenoxy)-2-methyl-3-[[(3S)-1-propan-2-ylpiperidin-3-yl]methyl]quinazolin-4-one;hydrochloride
74889155
>98%
Videos