missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Yellow Color Code Counter Shutters

Designed for use with Eppendorf Pipettes

Brand:  Eppendorf™ 3120815007

Code : NEW

Additional Details : Weight : 0.01100kg

 View more versions of this product

Product Code. 15224356

  • £3.96 / Each


Estimated Shipment
05-07-2018
Add to basket

Description

Description

  • Accessory for Eppendorf pipettes
  • For use with liquid handling applications
  • Counter shutters available for fixed and variable pipettes
Specifications

Specifications

For 100/200μL Input -channel Variable Pipette
1Each
Color Code Counter Shutter
Videos