missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Yeast Lysis Solution, for DNA isolation

Brand:  Alfa Aesar™ J61459.AK

Code : NEW

Additional Details : Weight : 0.00025kg

 View more versions of this product

Product Code. 15454059

  • £23.21 / 250mL


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Specifications

Colorless
DNA Extraction Reagent
2° to 8°C
Viscous Liquid
250mL
Videos
SDS