missing translation for 'onlineSavingsMsg'
Learn More

YC 1, Tocris Bioscience™

Soluble guanylyl cyclase (sGC) activator; induces G1 cell cycle arrest

Brand:  RD Systems (Bio-Techne) 4307/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13434534

  • £201.39 / 10mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Nitric oxide-independent activator of soluble guanylyl cyclase (sGC). Significantly elevates cGMP levels and inhibits collagen-stimulated aggregation of washed rabbit platelets (IC50 = 14.6 μM); induces relaxation in denuded phenylephrine-contracted rabbit aortic rings (EC50 = 1.9 μM). Also displays antiproliferative activity in vitro and in vivo by inducing G1 cell cycle arrest in two human hepatocellular carcinoma (HCC) cell lines, and in HCC xenografts in athymic SCID mice. Exhibits low cytotoxicity in non-malignant cells.
Specifications

Chemical Identifiers

170632-47-0
304.349
yc-1, lificiguat, yc 1, 3-5'-hydroxymethyl-2'-furyl-1-benzylindazole, unii-515cc1wpte, 515cc1wpte, 3-5'-hydroxymethyl-2'-furyl-1-benzyl indazole, 5-1-benzyl-1h-indazol-3-yl furan-2-yl methanol, 5-1-phenylmethyl-1h-indazol-3-yl-2-furanmethanol, 2-furanmethanol,5-1-phenylmethyl-1h-indazol-3-yl
[5-(1-benzylindazol-3-yl)furan-2-yl]methanol
C19H16N2O2
OQQVFCKUDYMWGV-UHFFFAOYSA-N
5712
C1=CC=C(C=C1)CN2C3=CC=CC=C3C(=N2)C4=CC=C(O4)CO

Specifications

10mg
170632-47-0
yc-1, lificiguat, yc 1, 3-5'-hydroxymethyl-2'-furyl-1-benzylindazole, unii-515cc1wpte, 515cc1wpte, 3-5'-hydroxymethyl-2'-furyl-1-benzyl indazole, 5-1-benzyl-1h-indazol-3-yl furan-2-yl methanol, 5-1-phenylmethyl-1h-indazol-3-yl-2-furanmethanol, 2-furanmethanol,5-1-phenylmethyl-1h-indazol-3-yl
C1=CC=C(C=C1)CN2C3=CC=CC=C3C(=N2)C4=CC=C(O4)CO
304.349
304.34
YC 1
C19H16N2O2
OQQVFCKUDYMWGV-UHFFFAOYSA-N
[5-(1-benzylindazol-3-yl)furan-2-yl]methanol
5712
>99%
Videos
Documents