Learn More
Y-27632 dihydrochloride, Tocris Bioscience™
Selective p160ROCK inhibitor
Brand:  Tocris (Bio-Techne) 125450
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13488983
  • £843.48 / 50mg
Estimated Shipment
04-10-2018
Add to basket
Description
Description
Selective p160ROCK inhibitor (Ki values are 0.14, 26, 25 and > 250 μM for p160ROCK, PKC, PKA and MLCK respectively). Also inhibits PRK2 (IC50 = 600 nM). Smooth muscle relaxant and orally active in vivo. Increases survival rate of human embryonic stem (hES) cells and iPSC-BMECs undergoing cryopreservation. Also optimizes naive human pluripotent stem cell growth and viability following naive cell derivation from primed ESCs and iPSCs using naive human stem cell medium (NHSM).
Specifications
Chemical Identifiers
129830-38-2
320.258
trans-4-[(1R)-1-Aminoethyl]-N-4-pyridinylcyclohexanecarboxamide dihydrochloride
4-[(1R)-1-aminoethyl]-N-pyridin-4-ylcyclohexane-1-carboxamide;dihydrochloride
C14H23Cl2N3O
IDDDVXIUIXWAGJ-DDSAHXNVSA-N
9901617
CC(C1CCC(CC1)C(=O)NC2=CC=NC=C2)N.Cl.Cl
Specifications
Y-27632 dihydrochloride
>98%
C14H23Cl2N3O
IDDDVXIUIXWAGJ-DDSAHXNVSA-N
4-[(1R)-1-aminoethyl]-N-pyridin-4-ylcyclohexane-1-carboxamide;dihydrochloride
9901617
129830-38-2
50mg
trans-4-[(1R)-1-Aminoethyl]-N-4-pyridinylcyclohexanecarboxamide dihydrochloride
CC(C1CCC(CC1)C(=O)NC2=CC=NC=C2)N.Cl.Cl
320.258
320.26
Videos