missing translation for 'onlineSavingsMsg'
Learn More

Y-26763, Tocris Bioscience™

Kir6 (KATP) channel opener

Brand:  Tocris (Bio-Techne) 207610

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13460424

  • £175.56 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Kir6 (KATP) channel opener and active metabolite of Y-27152 (Cat. No. 2077). Relaxes contracted rat aortic rings (IC50 = 0.027 mM) and inhibits glucose-induced insulin secretion in isolated human pancreatic β-cells in vitro. Produces hypotension in spontaneously hypertensive rats and displays cardioprotective properties in dogs following systemic administration in vivo.
Specifications

Chemical Identifiers

127408-31-5
276.292
unii-q9f3xv8a6x, q9f3xv8a6x, +-3s,4r-n-acetyl-n-hydroxy amino-6-cyano-3,4-dihydro-2,2-dimethyl-2h-1-benzopyran-3-ol, n-3s,4r-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-hydroxyacetamide, acetamide, n-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-hydroxy-, 3s-trans, n-3s,4r-6-cyano-3-hydroxy-2,2-dimethylchroman-4-yl-n-hydroxyacetamide, acetamide,n-3s,4r-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-hydroxy, n-3s,4r-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydro-1-benzopyran-4-yl-n-hydroxyacetamide, d05gpx, n-3s,4r-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydrochromen-4-yl-n-hydroxyacetamide
N-[(3S,4R)-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydrochromen-4-yl]-N-hydroxyacetamide
C14H16N2O4
IWTCFIIOUXJOOV-OLZOCXBDSA-N
121878
CC(=O)N(C1C(C(OC2=C1C=C(C=C2)C#N)(C)C)O)O

Specifications

Y-26763
127408-31-5
unii-q9f3xv8a6x, q9f3xv8a6x, +-3s,4r-n-acetyl-n-hydroxy amino-6-cyano-3,4-dihydro-2,2-dimethyl-2h-1-benzopyran-3-ol, n-3s,4r-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-hydroxyacetamide, acetamide, n-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-hydroxy-, 3s-trans, n-3s,4r-6-cyano-3-hydroxy-2,2-dimethylchroman-4-yl-n-hydroxyacetamide, acetamide,n-3s,4r-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2h-1-benzopyran-4-yl-n-hydroxy, n-3s,4r-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydro-1-benzopyran-4-yl-n-hydroxyacetamide, d05gpx, n-3s,4r-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydrochromen-4-yl-n-hydroxyacetamide
CC(=O)N(C1C(C(OC2=C1C=C(C=C2)C#N)(C)C)O)O
276.292
276.29
10mg
C14H16N2O4
IWTCFIIOUXJOOV-OLZOCXBDSA-N
N-[(3S,4R)-6-cyano-3-hydroxy-2,2-dimethyl-3,4-dihydrochromen-4-yl]-N-hydroxyacetamide
121878
>98%
Videos