missing translation for 'onlineSavingsMsg'
Learn More

Y 11, Tocris Bioscience™

Potent and selective FAK inhibitor

Brand:  RD Systems (Bio-Techne) 4498/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13464844

  • £124.20 / 10mg


Estimated Shipment
29-03-2019
Add to basket

Description

Description

Potent inhibitor of focal adhesion kinase (FAK); prevents FAK autophosphorylation at the Y397 site (IC 50 ∽ 50 nM in an in vitro kinase assay). Displays selectivity for FAK over a panel of other kinases (concentration used in assay = 1 μM). Decreases cell viability in a number of cancer cell lines; blocks colony formation in SW620 and BT474 cell lines.
Specifications

Chemical Identifiers

1086639-59-9
265.155
1-2-hydroxyethyl-1,3,5,7-tetraazatricyclo 3.3.1.1 3 ,? decan-1-ium bromide, 1-2-hydroxyethyl-3,5,7-triaza-1-azaniatricyclo 3.3.1.13,7 decane bromide, 3,7-triaza-1-azoniatricyclo 3.3.1.13,7 decane, 1-2-hydroxyethyl-, bromide
C1N2CN3CN1C[N+](C2)(C3)CCO.[Br-]
C8H17BrN4O
HBPXFHNNLMCUPA-UHFFFAOYSA-M
24195918

Specifications

Y 11
1086639-59-9
1-2-hydroxyethyl-1,3,5,7-tetraazatricyclo 3.3.1.1 3 ,? decan-1-ium bromide, 1-2-hydroxyethyl-3,5,7-triaza-1-azaniatricyclo 3.3.1.13,7 decane bromide, 3,7-triaza-1-azoniatricyclo 3.3.1.13,7 decane, 1-2-hydroxyethyl-, bromide
C1N2CN3CN1C[N+](C2)(C3)CCO.[Br-]
24195918
>98%
10mg
C8H17BrN4O
HBPXFHNNLMCUPA-UHFFFAOYSA-M
265.155
265.15
Videos