missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xylitol, 99%

Catalog Number p-7023417
Click to view available options
Quantity:
100 g
500 g
87-99-0
C5H12O5
152.15
MFCD00064291,MFCD00064292
HEBKCHPVOIAQTA-UHFFFAOYNA-N
xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
6912
OCC(O)C(O)C(O)CO
87-99-0
C5H12O5
152.15
MFCD00064291,MFCD00064292
HEBKCHPVOIAQTA-UHFFFAOYNA-N
xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
6912
OCC(O)C(O)C(O)CO

It is employed as a polyol substrate for xylitol and sorbitol dehydrogenases and sorbitol dehydrogenase derived from xylose. It is applied as a sweetener and humectants and skin conditioning agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a polyol substrate for xylitol and sorbitol dehydrogenases and sorbitol dehydrogenase derived from xylose. It is applied as a sweetener and humectants and skin conditioning agent in cosmetics industry.

Solubility
Soluble in water (50 mg/ml), abs methanol (6 g/100ml), abs ethanol (1.2 g/100ml), alcohol (very slightly), and pyridine. Insoluble in ether.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Store under inert gas.
TRUSTED_SUSTAINABILITY
Melting Point 92°C to 96°C
Density 1.515 g/mL
Assay Percent Range 99%
Quantity 100 g
Beilstein 1720523
Sensitivity Hygroscopic
Merck Index 14,10085
Solubility Information Soluble in water (50mg/ml),abs methanol (6g/100ml),abs ethanol (1.2g/100ml),alcohol (very slightly),and pyridine. Insoluble in ether.
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material Xylitol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'