missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenol Orange, BAKER™, J.T. Baker™
Click to view available options
Quantity:
5 g
Packaging:
Wide Mouth Amber Glass Bottle
Specifications
Formula Weight | 760.6 |
Packaging | Wide Mouth Amber Glass Bottle |
Quantity | 5 g |
Chemical Name or Material | Xylenol Orange |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'