missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylenes, contains ethylbenzene, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
417.05 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Description
- For Microelectronic Use
Specifications
Formula Weight | 106.17 |
Grade | BAKER ANALYZED™ ACS Reagent |
Quantity | 4 L |
Flash Point | 29.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 0.86kg |
Chemical Name or Material | Xylenes, Contains ethylbenzene |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'