Xylenes, 98+%, for analysis, mixture of isomers , ACROS Organics™


Overview
Brand: GC Internal (Acros/Geel)

Manufacturer Part Number: 383940050

5LT Xylenes, 98+%, for analysis, mixture of isomer

UNSPSC: 12190000

Code: 50

Additional Details:
Additional Details: CAS Number: 1330-20-7 Weight: 4.38000kg Transport: UN number : 1307 Chem class : 1 Pack group : III


Quantity 5L

Product Code. 10689404

Quantity Price
1 £ 63.9 / 5 L
Estimated Shipment
02-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Xylenes
CAS 1330-20-7
CAS 100-41-4
Purity Grade For analysis
Assay Percent Range 98+%
Molecular Formula C8H10
Formula Weight 106.17
Synonym Dimethylbenzene

Specification

Formula Weight 106.17
Quantity 5L
Purity Grade For analysis
CAS 1330-20-7
Assay Percent Range 98+%
Molecular Formula C8H10
Chemical Name or Material Xylenes
Synonym Dimethylbenzene
CAS 100-41-4
Documentation