missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylene, Certified AR for Analysis, Fisher Chemical™
39.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
39.57 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
39.57 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Description
C8H10, CAS Number-1330-20-7, 2.5L, 136 deg.C, Colorless, 106.17g/mol, -34 deg.C, 77264, Amber glass bottle, Liquid, 8mbar at 20 deg.C, 0.6 mPaS at 20 deg.C
Specifications
Melting Point | -34°C |
Color | Colorless |
Boiling Point | 136°C |
Quantity | 1 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 8mbar at 20°C |
Viscosity | 0.6 mPaS at 20°C |
Formula Weight | 106.17g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'