missing translation for 'onlineSavingsMsg'
Learn More

XTT sodium salt, Thermo Scientific Chemicals

Catalog Number 15424269
Click to view available options
Quantity:
250 mg

 

Tetrazolium derivative useful in cancer research.XTT sodium salt is used in conjunction with phenazine methosulfate to screen human cancer cell lines. It is also used to study fungal cell damage. It is involved in testing antimicrobial susceptibility of staphylococci and in candida biofilm research. Further, it is used in an assay for cell viability and cell proliferation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrazolium derivative useful in cancer research.XTT sodium salt is used in conjunction with phenazine methosulfate to screen human cancer cell lines. It is also used to study fungal cell damage. It is involved in testing antimicrob
TRUSTED_SUSTAINABILITY
CAS 111072-31-2
Color Yellow
Molecular Formula C22H16N7NaO13S2
MDL Number MFCD00083517
Quantity 250 mg
Synonym xtt
Solubility Information Soluble in hot water at 2;5mg/ml;Soluble in water,dimethyl sulfoxide,dimethyl formamide and phosphate buffered saline;at pH =7;2.
InChI Key JACYMBNQPPWQML-UHFFFAOYSA-M
SMILES [Na+].COC1=C(C=C(C(=C1)[N+]([O-])=O)S([O-])(=O)=O)N1N=C(N=[N+]1C1=C(OC)C=C(C(=C1)S([O-])(=O)=O)[N+]([O-])=O)C(=O)NC1=CC=CC=C1
IUPAC Name sodium;2-(2-methoxy-4-nitro-5-sulfonatophenyl)-3-(2-methoxy-4-nitro-5-sulfophenyl)-N-phenyltetrazol-3-ium-5-carboximidate
Molecular Weight (g/mol) 673.52
PubChem CID 131668615
Formula Weight 673.52
Physical Form Powder
Chemical Name or Material XTT sodium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'