missing translation for 'onlineSavingsMsg'
Learn More

XRP44X, Tocris Bioscience™

Ras-Net pathway inhibitor

Brand:  Tocris (Bio-Techne) 310150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13442444

  • £617.40 / 50mg

Estimated Shipment date
from Supplier 18-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ras-Net (Elk-3) pathway inhibitor (IC50 = 10 - 20 nM). Indirectly inhibits Net phosphorylation upstream of Erk1/2 activation. Mediates G2-M cell cycle arrest, suppresses growth of multiple cell types (IC50 ∽ 2 nM), inhibits angiogenesis and acts as a microtubule depolymerizing agent in vitro.
Specifications

Chemical Identifiers

729605-21-4
380.876
xrp44x, 4-3-chlorophenyl piperazin-1-yl 3-methyl-1-phenyl-1h-pyrazol-5-yl methanone, 1-3-chlorophenyl-4-3-methyl-1-phenyl-1h-pyrazol-5-yl carbonyl piperazine, 4-3-chlorophenyl-1-piperazinyl 3-methyl-1-phenyl-1h-pyrazol-5-yl-methanone, 1-3-chlorophenyl-4-5-methyl-2-phenylpyrazole-3-carbonyl piperazine, acmc-20ejss, xrp44x hplc, 4-3-chlorophenyl piperazin-1-yl 5-methyl-2-phenyl-2h-pyrazol-3-yl-methanone, 4-3-chlorophenyl piperazin-1-yl 5-methyl-2-phenyl-2h-pyrazol-3-yl methanone
[4-(3-chlorophenyl)piperazin-1-yl]-(5-methyl-2-phenylpyrazol-3-yl)methanone
C21H21ClN4O
NPHPNBGYPKBDDB-UHFFFAOYSA-N
10339779
CC1=NN(C(=C1)C(=O)N2CCN(CC2)C3=CC(=CC=C3)Cl)C4=CC=CC=C4

Specifications

50mg
729605-21-4
xrp44x, 4-3-chlorophenyl piperazin-1-yl 3-methyl-1-phenyl-1h-pyrazol-5-yl methanone, 1-3-chlorophenyl-4-3-methyl-1-phenyl-1h-pyrazol-5-yl carbonyl piperazine, 4-3-chlorophenyl-1-piperazinyl 3-methyl-1-phenyl-1h-pyrazol-5-yl-methanone, 1-3-chlorophenyl-4-5-methyl-2-phenylpyrazole-3-carbonyl piperazine, acmc-20ejss, xrp44x hplc, 4-3-chlorophenyl piperazin-1-yl 5-methyl-2-phenyl-2h-pyrazol-3-yl-methanone, 4-3-chlorophenyl piperazin-1-yl 5-methyl-2-phenyl-2h-pyrazol-3-yl methanone
CC1=NN(C(=C1)C(=O)N2CCN(CC2)C3=CC(=CC=C3)Cl)C4=CC=CC=C4
380.876
380.87
Ras GTPase Inhibitors
C21H21ClN4O
NPHPNBGYPKBDDB-UHFFFAOYSA-N
[4-(3-chlorophenyl)piperazin-1-yl]-(5-methyl-2-phenylpyrazol-3-yl)methanone
10339779
>99%
Videos
Documents