missing translation for 'onlineSavingsMsg'
Learn More

XCT 790, Tocris Bioscience™

Selective ERRα inverse agonist

Brand:  Tocris (Bio-Techne) 392850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13453824

  • £710.01 / 50mg

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective ERRα inverse agonist (IC50 = ∽400 nM). Displays no antagonist activity at ERRγ or ERα at concentrations below 10 μM. Interferes with PPARγ coactivator-1α (PGC-1α)/ERRα-dependent signaling: inhibits PGC-1α induction of ERRα and MAO-B gene expression and downregulates the constitutive transcriptional activity of ERRα in numerous cell lines.
Specifications

Chemical Identifiers

725247-18-7
596.428
3-4-2,4-bis-trifluoromethylbenzyloxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl acrylamide, d0zb7z, xct hplc , solid, 2e-3-4-2,4-bis trifluoromethyl phenyl methoxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl prop-2-enamide, e-3-4-2,4-bis trifluoromethyl benzyl oxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl acrylamide, e-3-4-2, 4-bis trifluoromethyl phenyl methoxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl prop-2-enamide, e-3-4-2,4-bis trifluoromethyl phenyl methoxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl prop-2-enamide, 3-4-2,4-bis trifluoromethyl phenyl methoxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl prop-2-enamide, 3-4-2,4-bis-trifluoromethyl-benzyloxy-3-methoxy-phenyl-2-cyano-n-5-trifluoromethyl-1,3,4 thiadiazol-2-yl-acrylamide
CHEBI:79999
COC1=C(C=CC(=C1)C=C(C#N)C(=O)NC2=NN=C(S2)C(F)(F)F)OCC3=C(C=C(C=C3)C(F)(F)F)C(F)(F)F
C23H13F9N4O3S
HQFNFOOGGLSBBT-AWNIVKPZSA-N
6918788
(E)-3-[4-[[2,4-bis(trifluoromethyl)phenyl]methoxy]-3-methoxyphenyl]-2-cyano-N-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]prop-2-enamide

Specifications

50mg
725247-18-7
3-4-2,4-bis-trifluoromethylbenzyloxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl acrylamide, d0zb7z, xct hplc , solid, 2e-3-4-2,4-bis trifluoromethyl phenyl methoxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl prop-2-enamide, e-3-4-2,4-bis trifluoromethyl benzyl oxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl acrylamide, e-3-4-2, 4-bis trifluoromethyl phenyl methoxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl prop-2-enamide, e-3-4-2,4-bis trifluoromethyl phenyl methoxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl prop-2-enamide, 3-4-2,4-bis trifluoromethyl phenyl methoxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl prop-2-enamide, 3-4-2,4-bis-trifluoromethyl-benzyloxy-3-methoxy-phenyl-2-cyano-n-5-trifluoromethyl-1,3,4 thiadiazol-2-yl-acrylamide
COC1=C(C=CC(=C1)C=C(C#N)C(=O)NC2=NN=C(S2)C(F)(F)F)OCC3=C(C=C(C=C3)C(F)(F)F)C(F)(F)F
596.428
CHEBI:79999
>99%
XCT 790
C23H13F9N4O3S
HQFNFOOGGLSBBT-AWNIVKPZSA-N
(E)-3-[4-[[2,4-bis(trifluoromethyl)phenyl]methoxy]-3-methoxyphenyl]-2-cyano-N-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]prop-2-enamide
6918788
596.42
Videos
Documents