missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ XCL1 Recombinant Human Protein

Expressed from DNA sequence encoding human XCL1 (Met 1- Gly 114) fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 10672H08B50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15237025

  • £227.11 / 50µg


Estimated Shipment
13-06-2018
Add to basket

Description

Description

  • Lyophilized in 20mM, Tris 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80, 10% glycerol
Specifications

Specifications

Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
6375
>85% by SDS-PAGE
Human
<1 EU/μg
XCL1
Cytokines and Receptors
XCL1 Recombinant Human Protein
Lyophilized
11.6kd
50μg
Full Length, Recombinant
Chemokine (C Motif) Ligand 1
Assay Standard
Insect
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.