missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ XCL1 (Human) ELISA Kit

For the Quantitative detection of Human XCL1.

Brand:  Abnova™ KA4207.1Kit

Additional Details : Weight : 0.02000kg

Product Code. 16038615

  • £508.00 / 1 set


Estimated Shipment
09-06-2020
Add to basket

Description

Description

  • XCL1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human XCL1.
Specifications

Specifications

Human
6375
Store at 4°C for 6 months. Store at -20°C for 12 months. Avoid multiple freeze-thaw cycles.
XCL1
chemokine (C motif) ligand 1
Kit
Human
1 Kit
Videos
Documents