missing translation for 'onlineSavingsMsg'
Learn More

XCC, Tocris Bioscience™

Adenosine receptor antagonist

Brand:  Tocris (Bio-Techne) 422050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13434384

  • £558.60 / 50mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Adenosine receptor antagonist (Ki values are 42, 68 and 1130 nM for A1, A2B and A2A receptors).
Specifications

Chemical Identifiers

96865-83-7
386.408
chembl27041, xcc, 4-2,6-dioxo-1,3-dipropyl-2,3,6,9-tetrahydro-1h-purin-8-yl phenoxy acetic acid, 2-4-2,6-dioxo-1,3-dipropyl-7h-purin-8-yl phenoxy acetic acid, d04dvt, gtpl451, 1,3-dipropyl-8-4-carboxymethyloxy phenylxanthine, 8-4-carboxymethyloxy phenyl-1,3-dipropylxanthine, 1,3-dipropyl-8-4-carboxymethoxy phenyl xanthine, 8-4'-carboxymethyloxyphenyl-1,3-dipropyl xanthine
2-[4-(2,6-dioxo-1,3-dipropyl-7H-purin-8-yl)phenoxy]acetic acid
C19H22N4O5
QTMMGCYGCFXBFI-UHFFFAOYSA-N
126079
CCCN1C2=C(C(=O)N(C1=O)CCC)NC(=N2)C3=CC=C(C=C3)OCC(=O)O

Specifications

50mg
96865-83-7
chembl27041, xcc, 4-2,6-dioxo-1,3-dipropyl-2,3,6,9-tetrahydro-1h-purin-8-yl phenoxy acetic acid, 2-4-2,6-dioxo-1,3-dipropyl-7h-purin-8-yl phenoxy acetic acid, d04dvt, gtpl451, 1,3-dipropyl-8-4-carboxymethyloxy phenylxanthine, 8-4-carboxymethyloxy phenyl-1,3-dipropylxanthine, 1,3-dipropyl-8-4-carboxymethoxy phenyl xanthine, 8-4'-carboxymethyloxyphenyl-1,3-dipropyl xanthine
CCCN1C2=C(C(=O)N(C1=O)CCC)NC(=N2)C3=CC=C(C=C3)OCC(=O)O
386.408
386.4
XCC
C19H22N4O5
QTMMGCYGCFXBFI-UHFFFAOYSA-N
2-[4-(2,6-dioxo-1,3-dipropyl-7H-purin-8-yl)phenoxy]acetic acid
126079
>98%
Videos
Documents