Learn More
Waters Corporation XBridge BEH130 C18 Column
The XBridge BEH130 C18 Peptide Separation Technology (PST) columns are optimized and QC tested for peptide separations.
£802.50 - £1155.00
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 130 Å |
Quantity | 1/Ea. |
USP Type | L1 |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
13195179
|
Waters Corporation
186003563 |
2.1 mm | 50 mm | 3.5 μm |
£937.50
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13125199
|
Waters Corporation
186003577 |
2.1 mm | 250 mm | 5 μm |
£1125.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13165179
|
Waters Corporation
186003560 |
1 mm | 50 mm | 3.5 μm |
£900.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13175179
|
Waters Corporation
186003561 |
1 mm | 100 mm | 3.5 μm |
£1005.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13185179
|
Waters Corporation
186003562 |
1 mm | 150 mm | 3.5 μm |
£1072.50
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
10044016
|
Waters Corporation
186003564 |
2.1 mm | 100 mm | 3.5 μm |
£1005.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13115189
|
Waters Corporation
186003565 |
2.1 mm | 150 mm | 3.5 μm |
£1072.50
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13125189
|
Waters Corporation
186003566 |
2.1 mm | 250 mm | 3.5 μm |
£1155.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13135189
|
Waters Corporation
186003567 |
4.6 mm | 50 mm | 3.5 μm |
£937.50
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13145189
|
Waters Corporation
186003568 |
4.6 mm | 100 mm | 3.5 μm |
£1005.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
10758117
|
Waters Corporation
186003569 |
4.6 mm | 150 mm | 3.5 μm |
£1072.50
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
10565344
|
Waters Corporation
186003570 |
4.6 mm | 250 mm | 3.5 μm |
£1155.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13165189
|
Waters Corporation
186003571 |
1 mm | 50 mm | 5 μm |
£900.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13175189
|
Waters Corporation
186003572 |
1 mm | 100 mm | 5 μm |
£967.50
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13185189
|
Waters Corporation
186003573 |
1 mm | 150 mm | 5 μm |
£1035.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13195189
|
Waters Corporation
186003574 |
2.1 mm | 50 mm | 5 μm |
£900.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13105199
|
Waters Corporation
186003575 |
2.1 mm | 100 mm | 5 μm |
£967.50
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13115199
|
Waters Corporation
186003576 |
2.1 mm | 150 mm | 5 μm |
£1035.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13135199
|
Waters Corporation
186003578 |
4.6 mm | 50 mm | 5 μm |
£900.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13145199
|
Waters Corporation
186003579 |
4.6 mm | 100 mm | 5 μm |
£967.50
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13155199
|
Waters Corporation
186003580 |
4.6 mm | 150 mm | 5 μm |
£1035.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13165199
|
Waters Corporation
186003581 |
4.6 mm | 250 mm | 5 μm |
£1125.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13145259
|
Waters Corporation
186003648 |
4.6 mm | 50 mm | 10 μm |
£802.50
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13155259
|
Waters Corporation
186003649 |
4.6 mm | 100 mm | 10 μm |
£870.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13165259
|
Waters Corporation
186003650 |
4.6 mm | 150 mm | 10 μm |
£930.00
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
13175259
|
Waters Corporation
186003651 |
4.6 mm | 250 mm | 10 μm |
£1027.50
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||
Description
- The small-pore (130 Å) trifunctionally bonded BEH particle offers you the widest usable pH range, superior low pH stability, and ultra-low column bleed to assay samples for proteomics, protein characterization, and peptide synthesis.
Specifications
6000 psi (415 Bar) | |
130 Å | |
L1 | |
Column |
Spherical | |
1/Ea. | |
Yes | |
Peptides |