missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Xanomeline oxalate
Functionally selective M1 agonist
POA
Specifications
Purity | >98% |
---|---|
Product Type | Xanomeline oxalate |
Molecular Weight (g/mol) | 371.46 |
Inhibitors | Xanomeline oxalate |
Description
Functionally selective muscarinic M1 receptor agonist (EC50 values are 0.3, 5, 42, 52 and 92.5 nM at M1, M3, M5, M4 and M2 receptors respectively). Displays a complex pharmacological profile: reversible and wash-resistant binding, resulting in full agonist activity at M1; delayed wash-resistant partial agonist activity at M2; and delayed wash-resistant full agonist activity at M4. Exhibits antipsychotic activity, and improves cognitive deficits and behavioral disturbances in Alzheimer's disease and schizophrenia.Specifications
>98% | |
Xanomeline oxalate | |
371.46 | |
Xanomeline oxalate |
For Research Use Only