missing translation for 'onlineSavingsMsg'
Learn More

Xamoterol hemifumarate, Tocris Bioscience™

Selective β1 partial agonist

Brand:  Tocris (Bio-Techne) 095050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13448193

  • £649.74 / 50mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

β1-adrenoceptor-selective partial agonist (pA2 values are 7.4 - 7.8 and 5.2 - 6.2 at β1- and β2-adrenoceptors respectively).
Specifications

Chemical Identifiers

73210-73-8
794.856
xamoterol hemifumarate, corwin, xamoterol fumarate, xamoterol fumarate usan, fumaric acid; bis xamoterol, e-but-2-enedioic acid; n-2-2-hydroxy-3-4-hydroxyphenoxy propyl amino ethyl morpholine-4-carboxamide, xamoterol hemifumarate salt, ici 118,587 hemifumarate salt, 1-4-hydroxyphenoxy-3-2-4-morpholinocarboxamido ethylamino-2-propanol hemifumarate
(E)-but-2-enedioic acid;N-[2-[[2-hydroxy-3-(4-hydroxyphenoxy)propyl]amino]ethyl]morpholine-4-carboxamide
C36H54N6O14
QEDVGROSOZBGOZ-WXXKFALUSA-N
6440459
C1COCCN1C(=O)NCCNCC(COC2=CC=C(C=C2)O)O.C1COCCN1C(=O)NCCNCC(COC2=CC=C(C=C2)O)O.C(=CC(=O)O)C(=O)O

Specifications

50mg
73210-73-8
xamoterol hemifumarate, corwin, xamoterol fumarate, xamoterol fumarate usan, fumaric acid; bis xamoterol, e-but-2-enedioic acid; n-2-2-hydroxy-3-4-hydroxyphenoxy propyl amino ethyl morpholine-4-carboxamide, xamoterol hemifumarate salt, ici 118,587 hemifumarate salt, 1-4-hydroxyphenoxy-3-2-4-morpholinocarboxamido ethylamino-2-propanol hemifumarate
C1COCCN1C(=O)NCCNCC(COC2=CC=C(C=C2)O)O.C1COCCN1C(=O)NCCNCC(COC2=CC=C(C=C2)O)O.C(=CC(=O)O)C(=O)O
794.856
397.43
Xamoterol hemifumarate
C36H54N6O14
QEDVGROSOZBGOZ-WXXKFALUSA-N
(E)-but-2-enedioic acid;N-[2-[[2-hydroxy-3-(4-hydroxyphenoxy)propyl]amino]ethyl]morpholine-4-carboxamide
6440459
>97%
Videos
Documents