Learn More
Fisherbrand™ Top-Line Pipette Filter Tips
Eliminate sample and pipette contamination. Fisherbrand™ Top-Line Pipette Filter Tips prevent impurities from passing through micropore filter.
Brand:  Fisherbrand™
Code : NEW
Additional Details : Weight : 1.98900kg
 View more versions of this product
Product Code. 10691213
  • £154.76 / Pack of 960
In Stock
to see stock.
Add to basket
Description
Description
  • Filters are made of high-quality hydrophobic virgin polyethylene without additives
  • Filters block aerosols and liquids through micropore filter technology
  • Optimized cone geometry for a secure fit
  • Graduation marks on tip at standard volumes (except 0.5-10/20μl filter tip)
  • Conical tip outlet prevents drop formation at the outlet opening of tip
  • Tips manufactured from autoclavable, non-wettable polypropylene
  • Sterile packs available (beta irradiation)
Specifications
Specifications
100 to 1000 μL
Sterile
Autoclavable
Beta radiation
Yes
Standard Volumes
Filtered
Clear
86mm
Rack
Virgin Polypropylene