Learn More
Fisherbrand™ Top-Line Pipette Filter Tips
Eliminate sample and pipette contamination. Fisherbrand™ Top-Line Pipette Filter Tips prevent impurities from passing through micropore filter.
Brand:  Fisherbrand™
Code : NEW
Additional Details : Weight : 0.96000kg
 View more versions of this product
Product Code. 10580813
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Filters are made of high-quality hydrophobic virgin polyethylene without additives
  • Filters block aerosols and liquids through micropore filter technology
  • Optimized cone geometry for a secure fit
  • Graduation marks on tip at standard volumes (except 0.5-10/20µl filter tip)
  • Conical tip outlet prevents drop formation at the outlet opening of tip
  • Tips manufactured from autoclavable, non-wettable polypropylene
  • Sterile packs available (beta irradiation)
Specifications
Specifications
1 to 50 μL
Sterile
Autoclavable
CE, IVD
Rack
Virgin Polypropylene
Filtered
Clear
50.8mm
Beta radiation
Yes
Standard Volumes