missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ WypAll™ L20 Fabric Wipers
Used for general cleaning and drying tasks. Kimberly-Clark™ WypAll™ L20 Fabric Wipers are thick, highly absorbent layered fabric that cleans up fast without falling apart.
POA
Specifications
Product Type | L20 Wipers |
---|---|
Material | Fabric |
Color | White |
For Use With (Application) | Light, precise wiping-up of spills, grime, dust; polishing glass and smooth surfaces; cleaning and drying tools and hands; finally assembly wiping; wiping food preparation areas |
Description
- Ideal for light, precise wiping-up of spills, grime and dust; polishing glass and smooth surfaces; assembly wiping; wiping food preparation areas
- A limited life wiper using AIRFLEX™ base sheet technology
- Available in small perforated rolls in white or blue for use in busy areas
Specifications
L20 Wipers | |
Fabric | |
White | |
Light, precise wiping-up of spills, grime, dust; polishing glass and smooth surfaces; cleaning and drying tools and hands; finally assembly wiping; wiping food preparation areas |