missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ WYPALL™ X80 Cloths, BRAG Box

Repeat-use wiper with HYDROKNIT™ technology. Kimberly-Clark™ WYPALL™ X80 Cloths, BRAG Box provides controlled access to hygienically protected wipers when cleaning or wiping on the move.

Brand:  Kimberly-Clark™ 8379

Code : 02

Additional Details : Weight : 0.01000kg

Product Code. 12625576

  • £55.50 / Each


Estimated Shipment
10-05-2018
Add to basket

Description

Description

  • 1 BRAG Box = 160 wipes
  • Cloths can be rinsed and reused
  • HYDROKNIT™ technology provides high absorbancy, strength and durability (wet or dry)
  • Resists most solvents, reduces residue, allows fast cleaning and helps reduce costs.
  • Ideal for heavy-duty tasks in industrial areas: wiping machinery and parts, cleaning rough surfaces without breaking up, wiping away grease, oil, solvents, heavier soiling and prepping surfaces.
Specifications

Specifications

2.281kg (5.03 lbs.)
42.7 x 31.8 cm
160 Sheets/Box
Heavy-duty tasks in industrial areas; wiping machinery and parts; cleaning rough surfaces without breaking up; wiping away grease, oil, solvents, heavier soiling; prepping surfaces
HYDROKNIT™ Fabric
White
Reusable
31.8 cm
42.7 cm
Heavy-duty wiper
Videos