missing translation for 'onlineSavingsMsg'
Learn More

Kimberly Clark Wiper roll L10 380mm x 235mm small roll of 200 sheets, for CMC-714-500F dispenser

Brand:  Kimberly Clark 7104

Wiper roll L10 380mm x 235mm small roll of 200 sheets, for CMC-714-500F dispenser

Code : 02

Additional Details : Weight : 5.04012kg

Product Code. 12794376

  • £41.00 / 12 rolls


In Stock 9
Add to basket