missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ WYPALL™ Cloths, POP-UP Box

Provides controlled access to hygienically protected wipers. Kimberly-Clark™ WYPALL™ X50 Cloths, POP-UP Box contains repeat-use wipers manufactured from highly absorbant HYDROKNIT™ fiber technology.

Brand:  Kimberly-Clark™ 8355

Code : 02

Additional Details : Weight : 0.01000kg

Product Code. 12695636

  • £169.00 / Pack of 10


Estimated Shipment
15-06-2018
Add to basket

Description

Description

  • Supplied in 10 Dispenser Boxes x 176 Sheets = 1760 Sheets
  • Repeat-use wiper with highly absorbant HYDROKNIT™ technology
  • Strong and durable (wet or dry), resists most solvents, reduces residue, allows fast cleaning and helps reduce costs.
  • Ideal for light-duty clean-up of adhesive, oil and debris, applying lubricants, thinners and solvents, cleaning glass, cleaning hands and precision cleaning of intricate machinery and parts
  • For cleaning or wiping on the move and need fewer wipers than the BRAG™ Box contains.
Specifications

Specifications

High
31.70 x 23.1cm
23.1cm
31.7cm
Cloth wipers
White
10 boxes x 176 sheets
Cleaning
Synthetic and Cellulose Fiber
Videos