missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark™ WYPALL™ X60 Cloths, Centerfeed Roll

Dispenses individual wipers one at a time to reduce waste while maintaining hygiene, especially in busy food areas. Kimberly-Clark™ WYPALL™ X60 Cloths, Centerfeed Roll is a repeat-use wiper constructed of highly absorbant HYDROKNIT™ fiber technology.

Brand:  Kimberly-Clark™ 8380

Code : 02

Additional Details : Weight : 0.01000kg

Product Code. 12665566

  • £21.70 / 1 roll


Estimated Shipment
13-02-2018
Add to basket

Description

Description

  • Supplied in a centerfeed roll of 150 sheets
  • Cloths can be rinsed and reused
  • HYDROKNIT™ technology provides high absorbancy, strength and durability (wet or dry)
  • Resists most solvents, reduces residue, allows fast cleaning and helps reduce costs.
  • Ideal for light-duty clean-up of adhesive, oil, debris, applying lubricants, thinners and solvents, cleaning glass, precision cleaning of intricate machinery and parts, cleaning hands.
Specifications

Specifications

High
Blue
150/Pk.
Cleaning
Synthetic and Cellulose Fiber
1.05kg (2.315 lbs.)
42 x 24.50cm
24.5cm
42cm
Cloth wipers
Videos