Learn More
WY 45233 succinate, Tocris Bioscience™
Dual serotonin/noradrenalin re-uptake inhibitor
Brand:  Tocris (Bio-Techne) 419450
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13464324
  • £352.80 / 50mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Serotonin and noradrenalin reuptake inhibitor (SNRI); selective for human serotonin (SERT) and noradrenalin (NET) transporters against 96 other targets (Ki values are 40.2 and 558.4 nM for SERT and NET respectively). Inhibits [3H]5-HT and [3H]NE uptake (IC50 values are 47.3 and 531.3 nM). Salt form of the major active metabolite of venlafaxine (Cat. No. 2917).
Specifications
Chemical Identifiers
448904-47-0
381.469
unii-h9e1t0bi90 component
butanedioic acid;4-[(1S)-2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol
C20H31NO6
ORUUBRMVQCKYHB-XFULWGLBSA-N
76958757
CN(C)CC(C1=CC=C(C=C1)O)C2(CCCCC2)O.C(CC(=O)O)C(=O)O
Specifications
50mg
448904-47-0
unii-h9e1t0bi90 component
CN(C)CC(C1=CC=C(C=C1)O)C2(CCCCC2)O.C(CC(=O)O)C(=O)O
381.469
381.46
WY 45233 succinate
C20H31NO6
ORUUBRMVQCKYHB-XFULWGLBSA-N
butanedioic acid;4-[(1S)-2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol
76958757
>99%