Learn More
WWC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558211
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RSGERSTRWY NLLSYKYLKK QSRELKPVGV MAPASGPAST DAVSALLEQT AVELEKRQEG RSSTQTLEDS WRYEETSENE Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.
Specifications
WWC1 | |
Polyclonal | |
Unconjugated | |
WWC1 | |
WW and C2 domain containing 1, FLJ10865, FLJ23369, HBEBP3, HBEBP36, KIAA0869, KIBRA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23286 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IX03 | |
WWC1 | |
Recombit protein corresponding to Human WWC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only