Learn More
WS 3, Tocris Bioscience™
TRPM8 agonist; cooling agent
Brand:  Tocris (Bio-Techne) 292750
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13432064
  • £308.70 / 50mg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
Cooling agent that is an agonist at TRPM8 receptors (EC50 = 3.7 μM).
Specifications
Chemical Identifiers
39711-79-0
211.349
n-ethyl-p-menthane-3-carboxamide, ethyl menthane carboxamide, n-ethyl-2-isopropyl-5-methylcyclohexanecarboxamide, ws-3, n-ethyl-4-menthane-3-carboxamide, cyclohexanecarboxamide, n-ethyl-5-methyl-2-1-methylethyl, fema no. 3455, ws 3, n-ethyl-2-isopropyl-5-methylcyclohexane carboxamide
N-ethyl-5-methyl-2-propan-2-ylcyclohexane-1-carboxamide
C13H25NO
VUNOFAIHSALQQH-UHFFFAOYSA-N
62907
CCNC(=O)C1CC(CCC1C(C)C)C
Specifications
50mg
39711-79-0
n-ethyl-p-menthane-3-carboxamide, ethyl menthane carboxamide, n-ethyl-2-isopropyl-5-methylcyclohexanecarboxamide, ws-3, n-ethyl-4-menthane-3-carboxamide, cyclohexanecarboxamide, n-ethyl-5-methyl-2-1-methylethyl, fema no. 3455, ws 3, n-ethyl-2-isopropyl-5-methylcyclohexane carboxamide
CCNC(=O)C1CC(CCC1C(C)C)C
211.349
211.34
WS 3
C13H25NO
VUNOFAIHSALQQH-UHFFFAOYSA-N
N-ethyl-5-methyl-2-propan-2-ylcyclohexane-1-carboxamide
62907
>99%