missing translation for 'onlineSavingsMsg'
Learn More

WS 12, Tocris Bioscience™

TRPM8 agonist; cooling agent

Brand:  Tocris (Bio-Techne) 304010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13452284

  • £149.94 / 10mg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent TRPM8 agonist that acts as a cooling agent (EC50 = 193 nM).
Specifications

Chemical Identifiers

68489-09-8
289.419
1r*,2s*-n-4-methoxyphenyl-5-methyl-2-1-methylethyl cyclohexanecarboxamide, 1r,2s,5r-2-isopropyl-n-4-methoxyphenyl-5-methylcyclohexane-1-carboxamide, d04ngl, n-4-methoxyphenyl-p-menthanecarboxamide, 1r,2s,5r-2-isopropyl-n-4-methoxyphenyl-5-methylcyclohexanecarboxamide, 1r,2s,5r-n-4-methoxyphenyl-5-methyl-2-1-methylethyl cyclohexane-carboxamide, 1r,2s,5r-n-4-methoxyphenyl-5-methyl-2-propan-2-yl cyclohexane-1-carboxamide, cyclohexanecarboxamide,n-4-methoxyphenyl-5-methyl-2-1-methylethyl-, 1r,2s,5r
(1R,2S,5R)-N-(4-methoxyphenyl)-5-methyl-2-propan-2-ylcyclohexane-1-carboxamide
C18H27NO2
HNSGVPAAXJJOPQ-XOKHGSTOSA-N
11266244
CC1CCC(C(C1)C(=O)NC2=CC=C(C=C2)OC)C(C)C

Specifications

10mg
68489-09-8
1r*,2s*-n-4-methoxyphenyl-5-methyl-2-1-methylethyl cyclohexanecarboxamide, 1r,2s,5r-2-isopropyl-n-4-methoxyphenyl-5-methylcyclohexane-1-carboxamide, d04ngl, n-4-methoxyphenyl-p-menthanecarboxamide, 1r,2s,5r-2-isopropyl-n-4-methoxyphenyl-5-methylcyclohexanecarboxamide, 1r,2s,5r-n-4-methoxyphenyl-5-methyl-2-1-methylethyl cyclohexane-carboxamide, 1r,2s,5r-n-4-methoxyphenyl-5-methyl-2-propan-2-yl cyclohexane-1-carboxamide, cyclohexanecarboxamide,n-4-methoxyphenyl-5-methyl-2-1-methylethyl-, 1r,2s,5r
CC1CCC(C(C1)C(=O)NC2=CC=C(C=C2)OC)C(C)C
289.419
289.41
WS 12
C18H27NO2
HNSGVPAAXJJOPQ-XOKHGSTOSA-N
(1R,2S,5R)-N-(4-methoxyphenyl)-5-methyl-2-propan-2-ylcyclohexane-1-carboxamide
11266244
>99%
Videos
Documents