Learn More
WR 1065, Tocris Bioscience™
p53 activator. Also ROS scavenger
Brand:  Tocris (Bio-Techne) 335650
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13462874
  • £310.21 / 50mg
Estimated Shipment
30-10-2018
Add to basket
Description
Description
Cell-permeable reactive oxygen species scavenger that activates p53 through a JNK-dependent signaling pathway. Activates p21waf-1 and MDM2, represses transcription of Myc and thymidine kinase, and inhibits DNA topoisomerase IIα. Induces cell cycle arrest and exhibits cytoprotective activity towards normal cells but not cancer cells in vivo. Also exhibits broad spectrum antiviral activity in vitro.
Specifications
Chemical Identifiers
14653-77-1
207.157
wr 1065 hydrochloride, amifostine thiol dihydrochloride, unii-le9vv16szw, wr-1065 dihydrochloride, 2-3-aminopropyl amino ethanethiol dihydrochloride, 2-3-aminopropylamino ethanethiol dihydrochloride, le9vv16szw, 2-3-aminopropyl amino ethanethiol dihydrochloride, ethanethiol, 2-3-aminopropyl amino-, dihydrochloride, 2-3-aminopropyl amino ethanethiol 2hcl
2-(3-aminopropylamino)ethanethiol;dihydrochloride
C5H16Cl2N2S
XDRLRDHLCIFZIW-UHFFFAOYSA-N
146169
C(CN)CNCCS.Cl.Cl
Specifications
WR 1065
14653-77-1
wr 1065 hydrochloride, amifostine thiol dihydrochloride, unii-le9vv16szw, wr-1065 dihydrochloride, 2-3-aminopropyl amino ethanethiol dihydrochloride, 2-3-aminopropylamino ethanethiol dihydrochloride, le9vv16szw, 2-3-aminopropyl amino ethanethiol dihydrochloride, ethanethiol, 2-3-aminopropyl amino-, dihydrochloride, 2-3-aminopropyl amino ethanethiol 2hcl
C(CN)CNCCS.Cl.Cl
207.157
207.16
50mg
C5H16Cl2N2S
XDRLRDHLCIFZIW-UHFFFAOYSA-N
2-(3-aminopropylamino)ethanethiol;dihydrochloride
146169
>98%
Videos