Learn More
WR 1065, Tocris Bioscience™
p53 activator. Also ROS scavenger
Brand:  Tocris (Bio-Techne) 335610
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13452874
  • £113.19 / 10mg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
Cell-permeable reactive oxygen species scavenger that activates p53 through a JNK-dependent signaling pathway. Activates p21waf-1 and MDM2, represses transcription of Myc and thymidine kinase, and inhibits DNA topoisomerase IIα. Induces cell cycle arrest and exhibits cytoprotective activity towards normal cells but not cancer cells in vivo. Also exhibits broad spectrum antiviral activity in vitro.
Specifications
Chemical Identifiers
14653-77-1
207.157
wr 1065 hydrochloride, amifostine thiol dihydrochloride, unii-le9vv16szw, wr-1065 dihydrochloride, 2-3-aminopropyl amino ethanethiol dihydrochloride, 2-3-aminopropylamino ethanethiol dihydrochloride, le9vv16szw, ethanethiol, 2-3-aminopropyl amino-, dihydrochloride, 2-3-aminopropyl amino ethanethiol 2hcl
2-(3-aminopropylamino)ethanethiol;dihydrochloride
C5H16Cl2N2S
XDRLRDHLCIFZIW-UHFFFAOYSA-N
146169
C(CN)CNCCS.Cl.Cl
Specifications
10mg
14653-77-1
wr 1065 hydrochloride, amifostine thiol dihydrochloride, unii-le9vv16szw, wr-1065 dihydrochloride, 2-3-aminopropyl amino ethanethiol dihydrochloride, 2-3-aminopropylamino ethanethiol dihydrochloride, le9vv16szw, ethanethiol, 2-3-aminopropyl amino-, dihydrochloride, 2-3-aminopropyl amino ethanethiol 2hcl
C(CN)CNCCS.Cl.Cl
207.157
207.16
WR 1065
C5H16Cl2N2S
XDRLRDHLCIFZIW-UHFFFAOYSA-N
2-(3-aminopropylamino)ethanethiol;dihydrochloride
146169
>98%