missing translation for 'onlineSavingsMsg'
Learn More

Wortmannin, Tocris Bioscience™

Potent, irreversible inhibitor of PI 3-kinase. Also inhibitor of PLK1

Brand:  Tocris (Bio-Techne) 12325

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 5mg

 View more versions of this product

Product Code. 13468923

  • £345.45 / 5mg

Estimated Shipment date
from Supplier 26-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, selective, cell-permeable and irreversible inhibitor of phosphatidylinositol 3-kinase (PI 3-kinase) (IC50 = 2 - 4 nM). Also potently inhibits polo-like kinase 1 (PLK1) (IC50 = 5.8 nM).
Specifications

Chemical Identifiers

19545-26-7
428.437
wortmannin, wartmannin, antibiotic sl-2052, unii-xva4o219qw, xva4o219qw, pi 3-kinase inhibitor, 1alpha,11alpha-11-acetyloxy-1-methoxymethyl-2-oxaandrosta-5,8-dieno 6,5,4-bc furan-3,7,17-trione, 1r,3r,5s,9r,18s-18-methoxymethyl-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo 10.6.1.0^ 2,10 .0^ 5,9 .0^ 15,19 nonadeca-2 10 ,12 19 ,14-trien-3-yl acetate, 1s,6br,9as,11r,11br-9a,11b-dimethyl-1-methyloxy methyl-3,6,9-trioxo-1,6,6b,7,8,9,9a,10,11,11b-decahydro-3h-furo 4,3,2-de indeno 4,5-h isochromen-11-yl acetate, wortmanin
CHEBI:52289
C23H24O8
QDLHCMPXEPAAMD-QAIWCSMKSA-N
312145
CC(=O)OC1CC2(C(CCC2=O)C3=C1C4(C(OC(=O)C5=COC(=C54)C3=O)COC)C)C

Specifications

5mg
19545-26-7
wortmannin, wartmannin, antibiotic sl-2052, unii-xva4o219qw, xva4o219qw, pi 3-kinase inhibitor, 1alpha,11alpha-11-acetyloxy-1-methoxymethyl-2-oxaandrosta-5,8-dieno 6,5,4-bc furan-3,7,17-trione, 1r,3r,5s,9r,18s-18-methoxymethyl-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo 10.6.1.0^ 2,10 .0^ 5,9 .0^ 15,19 nonadeca-2 10 ,12 19 ,14-trien-3-yl acetate, 1s,6br,9as,11r,11br-9a,11b-dimethyl-1-methyloxy methyl-3,6,9-trioxo-1,6,6b,7,8,9,9a,10,11,11b-decahydro-3h-furo 4,3,2-de indeno 4,5-h isochromen-11-yl acetate, wortmanin
CC(=O)OC1CC2(C(CCC2=O)C3=C1C4(C(OC(=O)C5=COC(=C54)C3=O)COC)C)C
312145
428.44
Wortmannin
C23H24O8
QDLHCMPXEPAAMD-QAIWCSMKSA-N
428.437
CHEBI:52289
>98%
Videos
Documents